Thermostatic baths for viscosity measurement, clear-view thermostats, Proline
Leverantör: LAUDA
L001532 L001533 L001534 L001535 L001536 L001537 L001538 L001539 L001540 L001541 L001546 L001547 L001548 L001549 L001550 L001551 L001552 L001553 L001554 L001555 L001560 L001561 L001562 L001563 L001564 L001565 L001566 L001567 L001568 L001569
461-1226EA 76425 SEK
461-1226 461-1228 461-1230 461-1227 461-1229 461-1231 461-1232 461-1234 461-1233 461-1235 LAUDL001546 LAUDL001547 LAUDL001548 LAUDL001549 LAUDL001550 LAUDL001551 LAUDL001552 LAUDL001553 LAUDL001554 LAUDL001555 LAUDL001560 LAUDL001561 LAUDL001562 LAUDL001563 LAUDL001564 LAUDL001565 LAUDL001566 LAUDL001567 LAUDL001568 LAUDL001569
Thermostatic baths for viscosity measurement, clear-view thermostats, Proline
Bad Bad med viskometer
The Clear-view thermostats are designed for direct observation of objects under temperature control and are thus particularly suitable for viscosity measurement.
  • Large front and back window panels
  • Choice of models to suit different ranges of temperature requirements - all with consistent temperature stability
  • Bath vessel made of stainless steel with handles and drain tap
  • Choice of thermostat, either Master or Command versions
The viscosity of Newtonian liquids is extremely dependent on temperature. Depending on the measurement medium, a deviation of 0,More Product Information