Heating thermostats, Proline
Leverantör: LAUDA
L001496 L001497 L001498 L001499 L001500 L001501 L001502 L001503 L001504 L001505 L001506 L001507 L001508 L001509 L001489
LAUDL001496EA 37400 SEK
LAUDL001496 LAUDL001497 LAUDL001498 LAUDL001499 LAUDL001500 LAUDL001501 LAUDL001502 LAUDL001503 LAUDL001504 LAUDL001505 LAUDL001506 LAUDL001507 LAUDL001508 LAUDL001509 461-1219
Heating thermostats, Proline
Termostater Termostatbad, öppna system Bad med Inbyggda Termostater
The Proline Edition X range of heating thermostats feature a wide temperature range, high cooling and heating power and a multi level pump. The self-explanatory menu-guided remote control Command is particularly convenient for operating the units at distances as far as 50 meters away. Thanks to the large display, the user can keep an eye on all the important parameters. There is a graphical display of measured values, large choice of interfaces (e.g. Ether- CAT, Ethernet, RS232/RS485, analogue module, Profibus, contact modules). Data collection together with documentation and control is easyMore Product Information