Process Thermostats, Variocool
Leverantör: LAUDA
L000983 L000985 L000656 L000657 L000658 L000659 L000660 L000661 L000663 L000664 L000665 L000668 L000669 L000670 L000671 L000672 L000673 L000674 L000675 L000677 L000678 L000728 L000729 L000730 L000746 L000747 L000748 L000949 L000951 L000953
LAUDL000983EA 160700 SEK
LAUDL000983 LAUDL000985 461-1307 461-1308 461-1309 461-1311 LAUDL000660 LAUDL000661 LAUDL000663 LAUDL000664 LAUDL000665 461-1318 461-1319 461-1323 461-1312 461-1313 461-1314 LAUDL000674 LAUDL000675 LAUDL000677 LAUDL000678 LAUDL000728 LAUDL000729 LAUDL000730 LAUDL000746 LAUDL000747 LAUDL000748 LAUDL000949 LAUDL000951 LAUDL000953
Process Thermostats, Variocool
Termostater Termostatbad, slutna system Cirkulationskylare
The compact Variocool circulating chillers are ideal for demanding temperature control tasks in laboratories, mini plants and production. The units have a colour TFT screen and membrane keyboard, a USB interface and an integrated alarm which are easily accessable on the front of the unit. They also feature an electronic fill gauge on the display and low fluid level alarm. Models can be either air or water cooled, all have lockable castors. A wide range of factory fitted options and interfaces which can be retrofitted give the user greater flexibility and control. Options include high powerMore Product Information