Polysorbat 80

Leverantör: Thermo Fisher Scientific

Synonyms: Sorbitanmonooleat, etoxylerad

L13315.AE L13315.AP L13315.0F
L13315.AEEA 715 SEK
L13315.AE L13315.AP L13315.0F
Polysorbat 80
Polysorbat 80

CAS No.: 9005-65-6 Flash Point: 149°(300°F) Refractive Index: 1.4750 Boiling Point: 100° Merck Reference: 14,7582 MDL No.: MFCD00082107

Kokpunkt: >100 °C
Smältpunkt: 100
Densitet: 1.064
Flampunkt: >149 °C (300 °F)
Storage Temperature: Omgivande
MDL Number: MFCD00082107
CAS nummer: 9005-65-6
EINECS: 500-019-9
Merck Index: 14,7582

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR