Acridine 97

Leverantör: Thermo Fisher Scientific
Varning

L01657.06 L01657.14
L01657.06EA 930 SEK
L01657.06 L01657.14
Acridine 97
Acridine

Purity: 98+% CAS No.: 260-94-6 Molecular Formula: C13H9N Formula Weight: 179.22 Melting Point: 107-111° Boiling Point: 346° Merck Reference: 14,122 UN No.: 2713 MDL No.: MFCD00005025

Formel: C₁₃H₉N
MW: 179.22 g/mol
Kokpunkt: 346 °C
Smältpunkt: 107…111 °C
Densitet: 1.005
Storage Temperature: Omgivande
MDL Number: MFCD00005025
CAS nummer: 260-94-6
EINECS: 205-971-6
UN: 2713
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR