Dispersing aggregates for POLYTRON® system PT10-35 GT
Leverantör: KINEMATICA
11030301 11030305 11030335 11030338 11030304 11030298 11030317 11030296 11030309 11030319
KINE11030301EA 28450 SEK
KINE11030301 KINE11030305 KINE11030335 KINE11030338 KINE11030304 KINE11030298 KINE11030317 KINE11030296 KINE11030309 KINE11030319
Dispersing aggregates for POLYTRON® system PT10-35 GT
Homogenisatorer Knivaggregat
Dispersing aggregates available as standard (EC design) and special aggregates (W, B design).
  • Manufactured from high-alloy stainless steel 316L, electrolytically polished
  • Simple removal and fast cleaning either by hand, dishwasher or autoclave
  • Easy disassembly of components
  • Fitted with a high-precision quick-release system
Standard aggregates: Useful in the preparation of samples and formulation development. The cutting teeth embedded in the head of the tool optimise the pre-crushing of the sample. An enclosed ring design provides optimum protectionMore Product Information