Sleeves for conical joints
Leverantör: KARTELL
1040 1035 1036 1037 1038 1039 1041 1042 1043
KART1040EA 750 SEK
KART1040 KART1035 KART1036 KART1037 KART1038 KART1039 KART1041 KART1042 KART1043
Sleeves for conical joints
Hylsor med NS-slipning NS-Manschetter
PTFE.
  • Very good chemical resistance
  • Gas, liquid and vacuum-tight (to 0,1 mbar)
  • Temperature resistant from −200 to +280 °C
For fitting onto standard ground cones. Easily detachable and suitable for repeated use. Joint grease is not needed.