α-Ketoglutaric acid disodium salt hydrate, Supelco®

Leverantör: Merck

Synonyms: Sodium 2-oxoglutarate dibasic hydrate, 2-Oxopentanedioic acid disodium salt hydrate

K3752-1KG K3752-100G K3752-5G
K3752-1KGEA 24080 SEK
K3752-1KG K3752-100G K3752-5G
α-Ketoglutaric acid disodium salt hydrate, Supelco®
α-Ketoglutaric acid disodium salt hydrate
Formel: C₅H₄Na₂O₅·xH₂O
MW: 190.06 g/mol (anhy)
MDL Number: MFCD00150702
CAS nummer: 305-72-6
EINECS: 206-167-8
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=7f2be19366d243882d9d14e9e6b65ed4a5e611f2&vid=b5ffb35acda6f2149e50d56614732609020b5c11&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=b1254b3e279fdf7651d6cfb9b728989a02a09d55&vid=2d1c2696daf65b58d6fe95d9f0668829c587d82f&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=afc76a7e4df012014050f99a262e6a328e505003&vid=f16c6522aa01ac4045ffad7f228f44db81b3a2a8&prid=web&clid=SAPDAM

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR