Spectrophotometer, 6300
Leverantör: Cole-Parmer
630531 83054-07
JENW630531EA 36075 SEK
JENW630531 JENW83054-07
Spectrophotometer, 6300
Spektrofotometer och fotometrer UV/Vis Spektrofotometrar
The 6300 is designed to provide a compact, high quality, easy to use measurement system.
  • Large sample compartment for a wide range of sampling devices to be fitted
  • Simple keypad layout and menu operating protocols
  • Display simultaneously shows both wavelength and the selected photometric value
  • Absorbance, transmittance and concentration measurement modes
Suitable for routine analysis in schools and colleges, as well as in QC and testing procedures for a wide variety of services and industries.
The 6320D model is based on the 6300 visibleMore Product Information