NP-40 lysis buffer

Leverantör: Thermo Fisher Scientific

J60766.AP J60766.AK
J60766.APEA 2375 SEK
J60766.AP J60766.AK
NP-40 lysis buffer
Buffertar Extraktionsbuffertar

Storage Temperature: 2 to 8 °C
Physical Form: Liquid
Appearance: Colourless

Order Now

SPECIFIKATIONER

Environmentally Preferable
Utseende Colourless
Fysikalisk form Liquid
Förvaringstemperatur 2°C to 8°C

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR