Laemmli SDS sample buffer, non-reducing (6X)

Leverantör: Thermo Fisher Scientific

J60660.ACEA 2125 SEK
J60660.AC J60660.AD
Laemmli SDS sample buffer, non-reducing (6X)
Buffertar Buffertar för elektrofores Laddningsbuffert

Physical Form: Liquid
Appearance: Blue
Applications: For protein sample preparation to be used in the Laemmli SDS-PAGE system

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR