TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedicals
Leverantör: MP Biomedicals
092020026
ICNA092020026EA 3375 SEK
ICNA092020026
TCH® Defined Serum Replacement, 50X Concentrate, MP Biomedicals
Cellodlingsmedium
This serum replacement is effective in research situations such as studying the effects of growth factors or in the manufacturing of cell derived products.
  • A universal serum replacement that supports human and mammalian cells
  • Completely free of virus, mycoplasma and other infectious agents
  • Contains no growth factors, steroid hormones, extraneous proteins or phenol red
  • Protein origin - human; Drug Master on file with the FDA
TCH is a fortified, multipurpose serum replacement for long-term culturing of primarily human anchorage-dependent and suspension cells. Highly effective in the production of biologicals, as well as in culturing bacteria cell culture and other mammalian cells.