Daunorubicin hydrochloride, powder

Leverantör: MP Biomedicals

Synonyms: Daunorubicin HCl, Hydroxydaunorubicin Hydrochloride, (8S,10S)-8-Acetyl-10-(((2R,4S,5S,6S)-4-amino-5-hydroxy-6-methyltetrahydro-2H-pyran-2-yl)oxy)-6,8,11-trihydroxy-1-methoxy-7,8,9,10-tetrahydrotetracene-5,12-dione hydrochloride

0215077705 0215077701
ICNA0215077705EA 1550 SEK
ICNA0215077705 ICNA0215077701
Daunorubicin hydrochloride, powder
Daunorubicin hydrochloride

Soluble in water, methanol, aqueous alcohols. Practically insoluble in chloroform, ether, benzene.


LD50 in mice 26 mg/kg in vitro.

Formel: C₂₇H₂₉NO₁₁·HCl
MW: 563.98 g/mol
MDL Number: MFCD04974507
CAS nummer: 23541-50-6

Order Now

Specifikation testresultat

Form Powder
Storage Store at +4 °C, protect from light.
Source Streptomyces peucetius


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR