TOC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Leverantör: Thermo Fisher Scientific
6PK40TOC10 6PK40TOC20
HYPE6PK40TOC10EA 8550 SEK
HYPE6PK40TOC10 HYPE6PK40TOC20
TOC certified screw vial and cap kits, level 3 high performance applications, SureSTART™
Vialer
Ensure that your environmental analyses meet government regulations with our level 3 Thermo Scientific™ SureSTART™ TOC certified screw vial and cap kits.
Select Level 3 products for ultimate performance and sensitivity from your methods, while maintaining the highest levels of sample security and integrity. Our TOC certified kits include 40 ml glass screw vials and pre-assembled polypropylene screw with bonded, high purity silicone/PTFE septa. Each lot of TOC vials is tested and certified to contribute background Total Organic Carbon levels of <10 or <20 ppb.