Crimp caps, 11 mm, level 3 high performance applications, SureSTART™
Leverantör: Thermo Fisher Scientific
6PCC11ST101 6PCC11TST 6PCCMST101X 6PCC11ST101X
HYPE6PCC11ST101EA 1075 SEK
HYPE6PCC11ST101 HYPE6PCC11TST HYPE6PCCMST101X HYPE6PCC11ST101X
Crimp caps, 11 mm, level 3 high performance applications, SureSTART™
Lock Lock, kromatografivialer
Use Thermo Scientific™ SureSTART™ 11 mm crimp caps with crimp vials that have an 11 mm opening, including SureSTART 2 ml glass crimp vials and glass crimp microvials for <2 ml samples (Level 3). Select level 3 products for ultimate performance and sensitivity from methods, while maintaining the highest levels of sample security and integrity.
Choose from aluminium and magnetic steel caps with all-purpose silicone/PTFE septa of various hardness values designed to reduce autosampler needle issues. Use Thermo Scientific™ manual or electronic crimpers and decappers for easy application andMore Product Information