Glass high recovery screw top 1,7 ml microvials for <2 ml samples, level 2 high-throughput applications, SureSTART™
Leverantör: Thermo Fisher Scientific
6ASV9-V1 6ASV9-V1T
HYPE6ASV9-V1EA 2525 SEK
HYPE6ASV9-V1 HYPE6ASV9-V1T
Glass high recovery screw top 1,7 ml microvials for <2 ml samples, level 2 high-throughput applications, SureSTART™
Vialer
Choose Thermo Scientific™ SureSTART™ 1,7 ml high recovery glass screw top microvials to maximise the injection volume for <2 ml samples in high-throughput applications.
With dimensions of 12×32 mm, these microvials have an internal conical reservoir that provides a total usable volume of 1,5 ml and a residual volume of 4 µl. Performance level 2 vials ensure high quality data with an uninterrupted workflow in high-throughput applications using GC, HPLC/UHPLC, and single or triple quadrupole MS systems. Use level 2 vials for routine QA/QC applications, where the analyte levels areMore Product Information