Immersion thermostats
Leverantör: HUBER
2000.0023.01 2061.0001.01
461-1047EA 23750 SEK
461-1047 HUBE2061.0001.01
Immersion thermostats
Termostater Termostatbad, öppna system Termostat Styrenheter
Immersion thermostats form the basis of many different combinations with polycarbonate or stainless steel bath vessels. All models are fitted with a powerful pressure/suction pump and meet the requirements of Safety Class III (FL) for use with flammable fluids. Temperatures down to −30 °C can be achieved in combination with a refrigerated bath. A screw clamp is included allowing the thermostats to be easily mounted on any vessels. A pump adapter for external temperature control and cooling coils for a cooling water connection are available as optional accessories.
  • Pilot ONE® is
More Product Information