Disposable particle filtering respirators, FFP2, OP-Air Pro®
Leverantör: Medicom
M52210-WH50
HOPEM52210-WH50EA 2025 SEK
HOPEM52210-WH50
Disposable particle filtering respirators, FFP2, OP-Air Pro®
Masker Andningsskydd, engångs Andningsskydd, partiklar
Designed to protect against high risk infectious diseases, these filtering half masks provide FFP2 NR D class protection against solid and liquid aerosols.
  • Category III and Medical Device Class I PPE
  • Bacterial Filtration Efficiency (BFE) is greater than 98%
  • Single-use, non sterile
  • Ultrasonic welding
  • Synthetic elastic headband
  • Latex-free
These folded masks feature a spunbond polypropylene inner/outer layer, with a melt-blown polypropylene filter. A polypropylene and iron nosepiece allows secure fitting. The masks provide 8 hours of effective protectionMore Product Information