Ammoniumdivätefosfat 100 g/L nitric acid 1% Matrix_Modifier_Solution, AAS, Fluka™

Leverantör: Honeywell Chemicals

Synonyms: Ammonium phosphate, monobasic, Ammonium Phosphate Monobasic, Ammonium dihydrogenphosphate

E4606-50ML
HONCE4606-50MLEA 3675 SEK
HONCE4606-50ML
Ammoniumdivätefosfat 100 g/L nitric acid 1% Matrix_Modifier_Solution, AAS, Fluka™
Ammoniumdivätefosfat

Certifikat: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB)

Formel: NH₄H₂PO₄
MW: 115.03 g/mol
Smältpunkt: 190
Densitet: 1.01 g/cm3
Storage Temperature: Omgivande
MDL Number: MFCD00003396
CAS nummer: 7722-76-1
Merck Index: 13,00547

Order Now

Specifikation testresultat

Matrix HNO₃ 1%
Concentration NH₄H₂PO₄ 100 g/L
Quality ISO/IEC17025:2017(ANAB),ISO17034:2016(ANAB)
Form Solution
Density 1.01g/cm3


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR