Magnesium AAS standards, (CRMs), Fluka™
Leverantör: Honeywell Chemicals
E4042-100ML E4042-500ML E4043-100ML E4043-500ML
HONCE4042-100MLEA 390 SEK
HONCE4042-100ML HONCE4042-500ML HONCE4043-100ML HONCE4043-500ML
Magnesium AAS standards, (CRMs), Fluka™
Standarder Cationic Standards Magnesium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifikat: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).