Oxalate standard for ion chromatography, (CRM), Fluka™
Leverantör: Honeywell Chemicals
E3037-100ML
HONCE3037-100MLEA 625 SEK
HONCE3037-100ML
Oxalate standard for ion chromatography, (CRM), Fluka™
Standarder Anionic Standards Oxalate Standards
Certified Reference Material (CRM)
  • Traceable to SRM from NIST
Certifikat: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).