Instrument check standard 5 for ICP
Leverantör: Honeywell Chemicals
E2111-100ML
HONCE2111-100MLEA 3800 SEK
HONCE2111-100ML
Instrument check standard 5 for ICP
Standarder Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
10 mg/L each of Mo, Sn, Sr, Ti in HNO₃ 2%