Instrument check standard 3 for ICP
Leverantör: Honeywell Chemicals
E2109-100ML
HONCE2109-100MLEA 2100 SEK
HONCE2109-100ML
Instrument check standard 3 for ICP
Standarder Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
200 mg/L each of Ca, Fe, K, Mg, Na in HNO₃ 2%