Instrument Calibration standard 2 for ICP and HGA elements
Leverantör: Honeywell Chemicals
E2104-100ML
HONCE2104-100MLEA 4250 SEK
HONCE2104-100ML
Instrument Calibration standard 2 for ICP and HGA elements
Standarder Cationic Standards Multi-Element Standards
Certified Reference Material (CRM).
400 mg/L Ni, 200 mg/L Zn, 150 mg/L Mn, 100 mg/L each of Ag, Cr in HNO₃ 5%