Titanium ICP Standards, (CRMs), Fluka™
Leverantör: Honeywell Chemicals
E1063-100ML E1125-100ML E1190-100ML E1257-100ML
HONCE1063-100MLEA 885 SEK
HONCE1063-100ML HONCE1125-100ML HONCE1190-100ML HONCE1257-100ML
Titanium ICP Standards, (CRMs), Fluka™
Standarder Cationic Standards Titanium Standards
Certified Reference Materials (CRMs)
  • Traceable to SRM from NIST
Certifikat: ISO/IEC 17025:2017 (ANAB), ISO 17034:2016 (ANAB).