Blood sedimentation pipette
Leverantör: GLASWARENFABRIK KARL HECHT
40382010
HECH40382010EA 735 SEK
HECH40382010
Blood sedimentation pipette
Pipetter Pipetter, hemometer
Blood sedimentation pipettes for the micro method.
  • Clear glass
  • Fused blue graduation “ASSISTENT-PERMANENT”