Reagent reservoir chillers
Leverantör: HEATHROW
120730 120731 120732 120733 120734 120735
HEAT120730EA 1625 SEK
HEAT120730 HEAT120731 HEAT120732 HEAT120733 HEAT120734 HEAT120735
Reagent reservoir chillers
Kylare Kylare, bänkmodell
These reagent reservoir chillers are the ice-free, non-toxic way to extend the working period and preserve integrity for temperature sensitive reagents. They are made from sonic welded, engineered PP that ensures a leak free design.
  • Reagents are kept at less than 5 °C for approximately 3 hours
  • Ice-free solution eliminates the risk of contamination from messy ice baths
  • Non-toxic blend of water and bacteriostatic agent chills and freezes quickly
  • Stackable, save space when storing in the freezer or on the bench top
  • Reusable, two-year warranty