BOD measurement system, BOD Direct Plus
Leverantör: Hach
LQV1669800001
HACHLQV1669800001EA 49775 SEK
HACHLQV1669800001
BOD measurement system, BOD Direct Plus
BOD mätsystem
BOD Direct Plus is the next generation BOD measuring instrument that delivers accurate, automatic and direct control of wastewater samples.
  • Large user friendly graphic display
  • Delivers free individual programming of each of the samples
  • Respirometric, electronic pressure sensor measuring principle
The inductive stirring system offers maintenance and wear free operation with automatic centring of stirring rods. With the built in automatic start function there is no possibility of pressure fluctuations related to the manometric measurement.