Conductivity standards
Leverantör: Hach
1440042 2307542 210542 2307442 1440049 2971849 1440026 LZW9700.99 LZW9710.99 LZW9720.99
HACH1440042EA 415 SEK
HACH1440042 HACH2307542 HACH210542 HACH2307442 HACH1440049 HACH2971849 HACH1440026 HACHLZW9700.99 HACHLZW9710.99 HACHLZW9720.99
Conductivity standards
Standarder Konduktivitetsstandarder
The sodium chloride standard solutions are used for calibrating conductivity meters and also used for accuracy checks.