Agaros 4% B - CL, cross-linked, beads

Leverantör: Thermo Fisher Scientific
Varning

H32305.AUEA 17950 SEK
H32305.AU
Agaros 4% B - CL, cross-linked, beads
Agaros

CAS No.: 61970-08-9 Storage Temperature: 2°C to 8°C Flash Point: 38°(100°F) Boiling Point: 100° UN No.: UN1170 MDL No.: MFCD00081294

Kokpunkt: 100 °C
Densitet: 1.040
Flampunkt: 38 °C (100 °F)
Storage Temperature: Kylskåp
MDL Number: MFCD00081294
CAS nummer: 9012-36-6
UN: 1170
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR