Cryoscope
Leverantör: Gerber Instruments AG
05.25501 05.25500
GEAG05.25501EA 0 SEK
GEAG05.25501 GEAG05.25500
Cryoscope
Fryspunktsmätare
The dynamic cooling system Cryoscope C1 is ideal for reliable and rapid determination of freezing point in milk acc. to ISO 5764/IDF 108. During the daily inspection of water content in the raw milk, the device works extremely quietly.
  • Easy to calibrate
  • Robust and compact design with touchscreen
  • Adjustable amplitude of the stirrer
  • Time or plateau mode selectable
  • Data output to an external printer and/or PC selectable
  • Worldwide support via internet possible
The dynamic cooling system allows a maximum sample throughput even at ambient temperatures up toMore Product Information