Ammoniumheptamolybdat tetrahydrat

Leverantör: FF CHEMICALS
Varning

Synonyms: Hexaammonium heptamolybdate tetrahydrate, Ammonium molybdate(VI) tetrahydrate, Ammonium molybdate tetrahydrate, Hexammonium heptamolybdate 4-hydrate, Molybdic acid ammonium salt tetrahydrate

FF60960050
FFCHFF60960050EA 0 SEK
FFCHFF60960050
Ammoniumheptamolybdat tetrahydrat
Ammoniumheptamolybdat tetrahydrat
Formel: (NH₄)₆Mo₇O₂₄·4H₂O
MW: 1235.86 g/mol
Densitet: 2.498 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00167059
CAS nummer: 12054-85-2
EINECS: 234-722-4
Merck Index: 13,00536

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR