S@feMate TOTAL Microbiological Safety Cabinets, Class II
Leverantör: BioAir S.p.A.
LDT4E0N BI00LDT4E00
EUCLLDT4E0NEA 0 SEK
EUCLLDT4E0N EUCLBI00LDT4E00
S@feMate TOTAL Microbiological Safety Cabinets, Class II
Dragskåp Biologiska säkerhetsbänkar
S@femate TOTAL Cabinets have been designed according to the airflow specifications for NSF49 Class II Type B2 cabinets, also known as 'total exhaust'. 100% fresh air is taken in from the lab, either from the front aperture and from a main inlet. The air from the main inlet is then cleaned by the main HEPA H14 filter to provide an ISO5 working area and joins with the air from the front aperture on the bottom of the cabinet. All the air is then circulated behind the backwall of the unit and filtered via the exhaust HEPA H14 filter and reinjected into the environment.
  • Sloped front sash (5°
More Product Information