AEC Peroxidase substrate kit
Leverantör: ENZO LIFE SCIENCES
ENZ43825
ENZOENZ43825EA 2175 SEK
ENZOENZ43825
AEC Peroxidase substrate kit
Analyser Enzymanalyser
The AEC Peroxidase Substrate Kit contains reagents for visualization of peroxidase reactions using Aminoethylcarbazole (AEC). In the presence of H2O2, peroxidases catalyze the oxidation of AEC into a brick red precipitate. The supplied reagents are sufficient for preparation of 300 ml of ready-to-use color reaction mixture.
Beställningsinformation: Contents: Aminoethylcarbazole, 6 ml 20 mg/ml in dimethylformamide 8X Reaction Buffer/Substrate Reagent, 40 ml Hydrogen peroxide in acetate buffer with stabilizers and enhancers Mixing Bottle, 1 15 ml bottle for preparation of color reaction mixture