Noladin ether 98 TLC_ANALYSMETOD

Leverantör: ENZO LIFE SCIENCES

BMLFA0330005
ENZOBMLFA0330005EA 2350 SEK
ENZOBMLFA0330005
Noladin ether 98 TLC_ANALYSMETOD
Noladin ether

Cannabinoid receptor ligand.


Endogenous cannabinoid agonist isolated from porcine brain. Binds specifically to CB₁ (Ki=21.2 nM) and CB₂ (Ki=>3 µM) receptors. Causes sedation hypothermia, intestinal immobility and mild antinociception in mice.


Soluble in DMSO (40 mg/ml) or ethanol (25 mg/ml).

Formel: C₂₃H₄₀O₃
MW: 364.6 g/mol
CAS nummer: 222723-55-9
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=53afbe2fbcb10eae9cc3e56fc00482bd38b908b8&vid=1dc2f8e3467c8b0d72a74418da3accd5c86e8259&prid=web&clid=SAPDAM

Order Now

Specifikation testresultat

Purity ≥98% (TLC)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR