CD40 Soluble (Human) ELISA kit
Leverantör: ENZO LIFE SCIENCES
ALX850262KI01
ENZOALX850262KI01EA 11450 SEK
ENZOALX850262KI01
CD40 Soluble (Human) ELISA kit
Analyser ELISAs
For the quantitative determination of human soluble CD40 in cell culture supernatants, serum and other body fluids.
  • Assay Time: <3,25 hours