Human recombinant TWEAK (soluble) (from E. coli)
Leverantör: ENZO LIFE SCIENCES
ALX522021C010
ENZOALX522021C010EA 6425 SEK
ENZOALX522021C010
Human recombinant TWEAK (soluble) (from E. coli)
Proteiner
Produced in E. coli. The extracellular domain of human TWEAK (aa 106-249) is fused at the N-terminus to a linker peptide (11 aa) and a FLAG®-tag.