Z-YVAD-pNA
Leverantör: ENZO LIFE SCIENCES
ALX260049M001
ENZOALX260049M001EA 590 SEK
ENZOALX260049M001
Z-YVAD-pNA
Proteiner
Caspase-1 substrate
Chromogenic substrate for caspase-1 with increased cell permeability. λmax of pNA is 400 nm.