Catalase fluorometric detection kit
Leverantör: ENZO LIFE SCIENCES
ADI907027
ENZOADI907027EA 5925 SEK
ENZOADI907027
Catalase fluorometric detection kit
Analyser Cellulär analys
The Catalase fluorometric detection kit provides a higher throughput sensitive method for measuring catalase activity in just one hour. This is a homogeneous mix-and-read assay that enables you to monitor multiple time points for a kinetic read. Positive control is provided.
  • Easily adaptable to HTS applications
  • Sensitive fluorescent detection system
For the measurement of catalase in cell lysates from any species.
Leveransinformation: Reaction buffer concentrate, Detection Reagent, Horseradish peroxidase concentrate, Hydrogen peroxide, Catalase enzyme