Substance P ELISA kit
Leverantör: ENZO LIFE SCIENCES
ADI900018A
ENZOADI900018AEA 6175 SEK
ENZOADI900018A
Substance P ELISA kit
Analyser ELISAs
Highly sensitive Substance P ELISA kit enabling detection of Substance P in plasma, serum, saliva, urine, and supernatants in just 3 hours.
  • Highly sensitive assay, measuring as little as 5.3 pg/ml of Substance P
  • Widely cited in numerous peer reviewed publications
  • High throughput fomat, assay up to 38 samples in duplicate per 96 well kit
  • Fully quantitative results that surpass semi-quantitative Western blot analysis
The Substance P EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 3 hours. Absorbance is read at 405 nm. This kit utilized low sample volume and provides the ability to assay up to 38 samples in duplicate per 96 well kit.
Beställningsinformation: Contains GxR IgG Microtiter plate, Conjugate, Antibody, Assay buffer, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution.
Observera: RUO - Research Use Only.