Trays, bare die/CSP, H 44 Series
Leverantör: ENTEGRIS
H44-740-62C02 H44-1550-141_U H4414017566C02 H44-0049-62C02 H44100150-1415 H44-750-1415 H4438619766C02 H4432850662C02 H4434820266C02 H4407814966C02 H44N61180366_U H44-600-66C02 H44-740-1415 H4478010566C02 H44-999-66C02 H44C1851301415 H44-499-1415 H44-600-1415_U H4410015062C02 H44500100-1415 H44-750-62C02 H44-999-62C02 H441952466C02 H44N50390362C2 H4449768666C02 H44-220-62C02 H4470281666C02 H44E24090162C2 H442802966C02 H44-1050-1415 H44-299-66C02 H44-400-1415 H44-600-62C02 H44-128-66C02 H44-218-66C02 H442503066C02 H44-355-66C02 H4435929266C02 H44-426-66C02 H4443954366C02 H445003066C02 H44-535-66C02 H4460675066C02 H44250300-1415 H4448183666C02 H44N22490566C2 H44-520-66C02 H4494531562C02 H44-999-74D10 H4425062566C02 H44E61355466C2 H4424426866C02 H44N40070066C2 H44E13013266C2 H44-740-66C02 H44C26641562C2 H44-999-74D05 H44-359-62C02 H4473465966C02 H44-0055-62C02 H44N01931266C0 H44-999-74D03 H44512282TW66C H44280540TW66C H44-999-1415 H44323299TW66C H44-0090-1403L H44266207TW66C
ENTEH44-740-62C02EA 0 SEK
ENTEH44-740-62C02 ENTEH44-1550-141_U ENTEH4414017566C02 ENTEH44-0049-62C02 ENTEH44100150-1415 ENTEH44-750-1415 ENTEH4438619766C02 ENTEH4432850662C02 ENTEH4434820266C02 ENTEH4407814966C02 ENTEH44N61180366_U ENTEH44-600-66C02 ENTEH44-740-1415 ENTEH4478010566C02 ENTEH44-999-66C02 ENTEH44C1851301415 ENTEH44-499-1415 ENTEH44-600-1415_U ENTEH4410015062C02 ENTEH44500100-1415 ENTEH44-750-62C02 ENTEH44-999-62C02 ENTEH441952466C02 ENTEH44N50390362C2 ENTEH4449768666C02 ENTEH44-220-62C02 ENTEH4470281666C02 ENTEH44E24090162C2 ENTEH442802966C02 ENTEH44-1050-1415 ENTEH44-299-66C02 ENTEH44-400-1415 ENTEH44-600-62C02 ENTEH44-128-66C02 ENTEH44-218-66C02 ENTEH442503066C02 ENTEH44-355-66C02 ENTEH4435929266C02 ENTEH44-426-66C02 ENTEH4443954366C02 ENTEH445003066C02 ENTEH44-535-66C02 ENTEH4460675066C02 ENTEH44250300-1415 ENTEH4448183666C02 ENTEH44N22490566C2 ENTEH44-520-66C02 ENTEH4494531562C02 ENTEH44-999-74D10 ENTEH4425062566C02 ENTEH44E61355466C2 ENTEH4424426866C02 ENTEH44N40070066C2 ENTEH44E13013266C2 ENTEH44-740-66C02 ENTEH44C26641562C2 ENTEH44-999-74D05 ENTEH44-359-62C02 ENTEH4473465966C02 ENTEH44-0055-62C02 ENTEH44N01931266C0 ENTEH44-999-74D03 ENTEH44512282TW66C ENTEH44280540TW66C ENTEH44-999-1415 ENTEH44323299TW66C ENTEH44-0090-1403L ENTEH44266207TW66C
Trays, bare die/CSP, H 44 Series
Waferhantering Brickor
Entegris offers a wide selection of H 44 series 4" square trays, covers and accessories to meet your changing needs. H 20 series trays are available in various pocket sizes and depths, with or without wash holes and tweezers slots. H 44 series products offer proven reliability for handling bare die, including:
  • Known good die (KGD)
  • Wafer scale packing (WSP)
  • Chip scal packing (CSP)
  • Flip chip
  • Bump die
  • Opto electronics
  • Passive and active chip components, mechanical components