ESD safety ankle boots, lace-up, Nature, JALAS® 6498
Leverantör: EJENDALS
6498-36 6498-37 6498-38 6498-39 6498-40 6498-41 6498-42 6498-43 6498-44 6498-45 6498-46 6498-47
EJEN6498-36EA 2482 SEK
EJEN6498-36 EJEN6498-37 EJEN6498-38 EJEN6498-39 EJEN6498-40 EJEN6498-41 EJEN6498-42 EJEN6498-43 EJEN6498-44 EJEN6498-45 EJEN6498-46 EJEN6498-47
ESD safety ankle boots, lace-up, Nature, JALAS® 6498
Skor och kängor
Anti-static ESD safety ankle shoes made from textile microfibre with EVA, polyester based conductive thread insole and Cambrelle® polyamide lining.
  • Double shock absorption zones in Poron® XRD®
  • ProNose toe reinforcement and oil resistant outsole
  • Composite toe protection cap
  • Puncture resistant midsole made from plasma-treated composite (PTC) textile
Mainly used for outdoors, all-year use, metal-free requirements for shoes and environments with risk for outsole penetration.
Colour: Black/grey/green
Certifikat: EN 61340-5-1 (ESD); EN ISO 20345:2011; S3, SRC