n-Tridekan

Leverantör: EHRENSTORFER
Fara

Synonyms: Tridecane

C17818000
EHERC17818000EA 1600 SEK
EHERC17818000
n-Tridekan
n-Tridekan
Formel: H₃C(CH₂)₁₁CH₃
MW: 184.37 g/mol
Kokpunkt: 235.4
Smältpunkt: –5.5 °C
Densitet: 0.7564 g/cm³ (20 °C)
Flampunkt: 79
Storage Temperature: Omgivande
MDL Number: MFCD00008979
CAS nummer: 629-50-5
EINECS: 211-093-4
UN: 3295
ADR: 3 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR