2,3,5,6-Tetrachloropyridine

Leverantör: EHRENSTORFER
Fara

C17376300
EHERC17376300EA 2925 SEK
EHERC17376300
2,3,5,6-Tetrachloropyridine
2,3,5,6-Tetrachloropyridine
Formel: C₅HCl₄N
MW: 216.88 g/mol
Kokpunkt: 251.6
Smältpunkt: 88…89 °C
MDL Number: MFCD00092428
CAS nummer: 2402-79-1
EINECS: 219-283-9
UN: 2811
ADR: 6.1 III

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR