2-Ftalsyra

Leverantör: EHRENSTORFER
Varning

Synonyms: Pathalic acid

C16167500
EHERC16167500EA 1550 SEK
EHERC16167500
2-Ftalsyra
2-Ftalsyra
Formel: C₈H₆O₄
MW: 166.13 g/mol
Kokpunkt: 315…317 °C (1013 hPa)
Smältpunkt: 79
Densitet: 1.576 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002467
CAS nummer: 88-99-3
EINECS: 201-873-2
Merck Index: 12,07527

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR