2-Pentadecanone

Leverantör: EHRENSTORFER

Synonyms: Pentadecan-2-one

C15973802
EHERC15973802EA 980 SEK
EHERC15973802
2-Pentadecanone
2-Pentadecanone
Formel: C₁₅H₃₀O
MW: 226.4 g/mol
Densitet: 0.82 g/cm³ (39 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00053712
CAS nummer: 2345-28-0
EINECS: 219-064-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR