(4-Chlorophenyl)acetic acid

Leverantör: EHRENSTORFER
Varning

Synonyms: 2-(4-Chlorophenyl)acetic acid, p-Chlorophenylacetic Acid

C11489000
EHERC11489000EA 1400 SEK
EHERC11489000
(4-Chlorophenyl)acetic acid
(4-Chlorophenyl)acetic acid
Formel: (C₆H₄Cl)CH₂COOH
MW: 170.6 g/mol
Kokpunkt: 292
Smältpunkt: 101…104 °C
Densitet: 1.43 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00004344
CAS nummer: 1878-66-6
EINECS: 217-521-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR