Benzamid

Leverantör: EHRENSTORFER
Varning

Synonyms: Benzoic acid amide

C10532250
EHERC10532250EA 1525 SEK
EHERC10532250
Benzamid
Benzamid
Formel: C₇H₇NO
MW: 121.14 g/mol
Kokpunkt: 288
Smältpunkt: 124
Densitet: 1.35 g/cm³ (20 °C)
Flampunkt: 180
Storage Temperature: Omgivande
MDL Number: MFCD00007968
CAS nummer: 55-21-0
EINECS: 200-227-7
Merck Index: 13,01059

Order Now

SPECIFIKATIONER

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR