p-Anisic acid

Leverantör: EHRENSTORFER

Synonyms: 4-Methoxybenzoic acid, 4-Anisic acid

C10265900
EHERC10265900EA 1125 SEK
EHERC10265900
p-Anisic acid
p-Anisic acid
Formel: C₈H₈O₃
MW: 152.15 g/mol
Kokpunkt: 275
Smältpunkt: 182…185 °C
Densitet: ∼1.385 g/cm³ (20 °C)
Storage Temperature: Omgivande
MDL Number: MFCD00002542
CAS nummer: 100-09-4
EINECS: 202-818-5
Merck Index: 13,00669

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR