Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate 99%, Sigma-Aldrich®

Leverantör: SIGMA ALDRICH MICROSCOPY
Varning

Synonyms: Ethyl trans-α-cyano-3-indoleacrylate

E18905-1G
E18905-1GEA 1850 SEK
E18905-1G
Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate 99%, Sigma-Aldrich®
Ethyl(2E)-2-cyano-3-(1H-indolyl-3-yl)acrylate

Example of a cyanovinylheteroaromatic for organic nonlinear optics.

Formel: C₁₄H₁₂N₂O₂
MW: 240.26 g/mol
Smältpunkt: 164...166 °C (dec., lit.)
Storage Temperature: Room Temperature
MDL Number: MFCD00022742
CAS nummer: 62309-96-0
UN: 3077
ADR: 9 III
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=8878b74c065810c198ba9f41910a25e0192bc64f&vid=8878b74c065810c198ba9f41910a25e0192bc64f&prid=web&clid=SAPDAM

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Läs mer About VWR